Learn More
Alfa Aesar™ Albumin, Bovine, New Zealand origin, 97%, Chromatographically Purified
For research use
Manufacturer:  Alfa Aesar™ J6491214
 View more versions of this product
Catalog No. AAJ6491214
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
Suitable for immunoassay applications where protease activity would be undesirable
Lyophilized powder
Bovine Serum
RUO
No
Lyophilized Powder
25g
2° to 8°C
Albumin, Bovine, New Zealand origin, 97%, Chromatographically purified