Learn More
Alfa Aesar™ Albumin, Bovine, New Zealand origin, Low Fatty Acid, Chromatographically Purified
For research use
Manufacturer:  Alfa Aesar™ J6540014
 View more versions of this product
Catalog No. AAJ6540014
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
Suitable for immunoassays. Reduced fatty acid content minimizes non-specific binding of antibodies
Lyophilized powder
Bovine Serum
RUO
No
Lyophilized Powder
25g
2° to 8°C
Albumin, Bovine, New Zealand origin, Low Fatty Acid, Chromatographically purified