Learn More
Albumin, Bovine Serum, Fraction V, Heat-Shock Isolation, BiotechGrade, Spectrum™
A3611, 9048-46-8
Manufacturer:  Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner A3611100GM
 View more versions of this product
Catalog No. 18-600-352
Add to cart
Description
Description
Spectrum™ Albumin, Bovine Serum, Fraction V, Heat-Shock Isolation, BiotechGrade is a serum albumin protein that comes from cows. It is also known as Bovine serum albumin and usually used as a protein concentration standard in laboratory experiments. Spectrum offers highly pure reagents suitable for biochemical research and analysis.
Specifications
Specifications
Lyophilized Powder
100 g
Poly Bottle