Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Albumin, Human, 96%

For research use

Manufacturer:  Alfa Aesar™ J6678006

 View more versions of this product

Catalog No. AAJ6678006


Order in Bulk

Request bulk or custom formats

Add to cart

Specifications

Specifications

Produced in the liver, albumin is the most abundant protein in blood plasma. A carrier protein for steroids, fatty acids, and thyroid hormones, and is vital in regulating the colloidal osmotic pressures of blood. Binds exogenous substances.
Lyophilized powder
5g
2° to 8°C
Albumin, Human, 96%
Lyophilized Powder
96%
Human Serum
RUO
No
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only