Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Albumin Monoclonal Antibody (C5-A8), Huabio
Mouse Monoclonal Antibody
Supplier: HUABIO RESEARCH INC EM190180


Description
Albumin Monoclonal antibody specifically detects Albumin in Human, Rat samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
Albumin | |
Monoclonal | |
2 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
P02768, P02770 | |
Mouse | |
Protein A | |
RUO | |
213, 24186 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
C5-A8 | |
Unconjugated | |
ALB | |
ALB; ALB protein; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; Bos d 6; bovine serum albumin; BSA; cell growth inhibiting protein 42; FDAH; GIG20; GIG42; growth-inhibiting protein 20; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341 | |
Native protein. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction