Learn More
Albumin Mouse, Alexa Fluor 700, Clone: HSA20, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP179049AF700
Catalog No. NBP179049AI
Add to cart
Description
Description
Albumin Monoclonal antibody specifically detects Antigen in Bovine (Negative), Human samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications
Specifications
Albumin
HSA20
Western Blot, ELISA
Purified
ALB
Human serum albumin
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
No cross-reactivity with other human proteins tested.
ELISA, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Mouse
IgG1
0.2 ml
Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers
Primary
213
Bovine (Negative), Human