Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albumin Mouse anti-Human, Alexa Fluor™ 350, Clone: 5B11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223495AF350

Catalog No. NP22349A30


Add to cart

Description

Description

Albumin Monoclonal antibody specifically detects Albumin in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

Albumin
5B11
Western Blot, ELISA
albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Clone 5B11 is specific for native and denatured human serum albumin (HSA)
ELISA, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
ALB
Native HSA isolated from human plasma adsorbed onto aluminum hydroxide gel
100 μL
Primary
213
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit