Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albumin, Mouse anti-Human, Clone: 188815, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $304.50

Specifications

Antigen Albumin
Clone 188815
Immunogen Human Serum Albumin
Host Species Mouse
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB14551100
View Documents
R&D Systems
MAB14551100
100 ug
Each for $304.50
Add to cart
 
MAB14551SP
View Documents
R&D Systems
MAB14551SP
25 ug
Each for $119.60
Add to cart
 
Description

Description

Albumin Monoclonal antibody specifically detects Albumin in Human Samples. It is validated for ELISA.
Specifications

Specifications

Albumin
Human Serum Albumin
Primary
Human
Unconjugated
213
Purified
ELISA
RUO
188815
Mouse
Monoclonal
IgG2b
ALB, albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit