Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albumin Mouse anti-Human, Clone: OTI3B10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270149

Catalog No. NBP270149


Add to cart

Description

Description

Albumin Monoclonal antibody specifically detects Albumin in Human samples. It is validated for Western Blot.
Specifications

Specifications

Albumin
OTI3B10
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Mouse
IgG1
Immunogen affinity purified
Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers
Primary
213
Human
Western Blot
LYOPH
Western Blot 1:2000
Affinity Purified, Lyophilized
ALB
Human recombinant protein fragment corresponding to amino acids 19-609 of human ALB (NP_000468) produced in E.coli.
67.2 kDa
100 μg
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit