Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albumin Mouse, Biotin, Clone: HSA20, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP179049B

Catalog No. NBP179049B


Add to cart

Description

Description

Albumin Monoclonal antibody specifically detects Antigen in Bovine (Negative), Human samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications

Specifications

Albumin
HSA20
Western Blot, ELISA
Purified
ALB
Human serum albumin
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
No cross-reactivity with other human proteins tested.
ELISA, Western Blot
Biotin
PBS with 0.05% Sodium Azide
albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Mouse
IgG1
0.2 ml
Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers
Primary
213
Bovine (Negative), Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit