Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Albumin Reagent
Determine in vitro quantity of albumin in serum or plasma.
Description
Determine the quantity of albumin in serum or plasma in vitro with Thermo Scientific Albumin Reagent.
Features:- Available in various configurations and kit sizes to suit small and large laboratory needs
- Suitable for manual use or on automated clinical chemistry analyzers
- Applications for a wide range of automated analyzers available on request
- Utilizes the Bromocresol Green substrate
Albumin is the most abundant protein in human plasma, representing 55 to 65% of the total protein. Albumin has a relatively small size which makes it a useful indicator of the integrity of glomerular and other membranes. Hyperalbumin is associated with dehydration while hypoalbuminemia is associated with many illnesses including, but not limited to, kidney disease, liver disease, tissue damage, inflammation and malnutrition.
Specifications
CE Marked | |
2 x 250mL | |
2°C to 8°C | |
Yes | |
Albumin, Quantitative determination of albumin in serum or plasma | |
Endpoint |
Liquid | |
Plasma, Serum | |
Reagent | |
Albumin - Bromocresol Green | |
0 to 60g/L | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title