Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albuterol Sulfate, USP, 98.5-101%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
51022-70-9
C26H44N2O10S
576.70
BNPSSFBOAGDEEL-UHFFFAOYNA-N
bis(4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol); sulfuric acid
OS(O)(=O)=O.CC(C)(C)NCC(O)C1=CC=C(O)C(CO)=C1.CC(C)(C)NCC(O)C1=CC=C(O)C(CO)=C1
51022-70-9
C26H44N2O10S
576.70
BNPSSFBOAGDEEL-UHFFFAOYNA-N
bis(4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol); sulfuric acid
OS(O)(=O)=O.CC(C)(C)NCC(O)C1=CC=C(O)C(CO)=C1.CC(C)(C)NCC(O)C1=CC=C(O)C(CO)=C1

AL156, 51022-70-9, (C13H21NO3)2•H2SO4

Spectrum™ Albuterol Sulfate, USP is a bronchodilator used to treat or prevent bronchospasm in people with reversible obstructive airway disease. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Quantity 1 kg
CAS Max % 100%
Percent Purity 98.5 to 101%