Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALCAM/CD166 Mouse anti-Human, Mouse, Allophycocyanin, Clone: 4H9A5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237345APC

Catalog No. NBP237345T

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ALCAM/CD166
4H9A5
Flow Cytometry
Purified
ALCAM
Purified recombinant fragment of human ALCAM (AA: 48-216) expressed in E. Coli.
Protein A or G purified
Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
214
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit