Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALCAM/CD166 Rabbit anti-Cynomolgus Monkey, DyLight 755, Clone: 8, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP290954IR

Catalog No. NB041421


Add to cart

Description

Description

ALCAM/CD166 Monoclonal antibody specifically detects ALCAM/CD166 in Cynomolgus Monkey samples. It is validated for Western Blot
Specifications

Specifications

ALCAM/CD166
8
Western Blot
Purified
Rabbit
IgG
0.1 mL
Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
214
Western Blot
DyLight 755
50mM Sodium Borate
activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733
Recombinant Cynomolgus ALCAM/CD166 protein
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Cynomolgus Monkey
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit