Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALCAM Mouse anti-Human, Clone: 3F8B12, Abnova™

Mouse monoclonal antibody raised against human ALCAM.

Manufacturer:  Abnova Corporation MAB12605

Catalog No. 89-933-545


Add to cart

Description

Description

Specifications

Specifications

ALCAM
3F8B12
Mouse monoclonal antibody raised against human ALCAM.
In ascites (0.03% sodium azide).
CD166/FLJ38514/MEMD/MGC71733
Mouse
IgG2a
RUO
Primary
214
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:10000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:400) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Liquid
ALCAM
Recombinant protein corresponding to amino acids 405-524 of human ALCAM.
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit