Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alclometasone Dipropionate, USP, 97-102%, Spectrum™

Click to view available options
Quantity:
50 g
100 g
Packaging:
Amber Glass Bottle
66734-13-2
66734-13-2

A1574, 66734-13-2, C28H37ClO7

Spectrum™ Alclometasone Dipropionate, USP is a low- to medium-strength corticosteroid. It is used to treat a variety of skin conditions (e.g., eczema, dermatitis, allergies, rash). Alclometasone reduces the swelling, itching, and redness that can occur in these types of conditions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 50 g
Specific Rotation +21° to +25°
CAS Max % 100%
Percent Purity 97 to 102%