Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol, 190 Proof, BiotechGrade, 95-96%, Spectrum™

Click to view available options
Quantity:
4 L
500 mL
Packaging:
Amber Glass Bottle
64-17-5
C2H6O
46.07
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
CCO
64-17-5
C2H6O
46.07
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
CCO

E1421, 64-17-5, C2H5OH

Spectrum™ Alcohol, 190 Proof, BiotechGrade is an extremely volatile, flammable, colorless liquid, pure ethyl alcohol (ethanol) that are ideal for applications in industrial, health, education, and research. Biotech grade is a reliable, high-quality, reagent for general laboratory use and is commonly used in thermometers, as fuel and as a powerful solvent. Spectrum offers highly pure reagents suitable for biochemical research and analysis that are absence of inhibitors.
CAS Max % 100%
UN Number UN1170
Percent Purity 95 to 96%
Packaging Amber Glass Bottle
Quantity 500 mL