Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol, 190 Proof, USP, 94.9-96%, Spectrum™

Click to view available options
Quantity:
1 L
100 mL
4 L
4 x 4 L
500 mL
55 gal.
6 x 1 L
6 x 500 mL
Packaging:
Amber Glass Bottle
Drum
64-17-5
C2H6O
46.07
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
CCO
64-17-5
C2H6O
46.07
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
CCO

ET108, 64-17-5, C2H5OH

Spectrum™ Alcohol, 190 Proof, USP is approximately 95% Ethanol and 5% water. This product, 190 proof USP grade alcohol, is used as an antiseptic to disinfect skin prior to injections and as a solvent. All Spectrum Chemical USP products, including all USP alcohol products, are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
UN Number UN1170
Benzene 2 ppm
Percent Purity 94.9 to 96%
Methanol 200 ppm
Non_volatile Matter 2.5 mg
Packaging Amber Glass Bottle
Specific Gravity 0.812-0.816
Quantity 6 x 1 L