Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle
Poly Bottle
64-17-5
CH4O
32.042
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit
702
CHEBI:16236
methanol
CO
64-17-5
CH4O
32.042
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit
702
CHEBI:16236
methanol
CO

High quality chemicals for laboratory and specialized industrial use

    For Histological Use; NOTE: Intended for nonfuel end purposes only
Grade Meets ACS reagent chemical requirements
Packaging Poly Bottle
Specific Gravity 1L = 0.79kg
Flash Point 13.00°C (55.4°F)
Quantity 4L
Chemical Name or Material Alcohol, Anhydrous