Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle
Poly Bottle

Chemical Identifiers

67-56-1
CH4O
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
CHEBI:17790

High quality chemicals for laboratory and specialized industrial use

    For Histological Use; NOTE: Intended for nonfuel end purposes only
SMILES CO
IUPAC Name methanol
Packaging Narrow Mouth PVC Coated Amber Glass Bottle
Specific Gravity 1L = 0.79kg
Flash Point 13.00°C (55.4°F)
Quantity 4L
Chemical Name or Material Alcohol, Anhydrous
Grade Baker Analyzed™ ACS Reagent, Meets ACS reagent chemical requirements