Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol, Anhydrous, Denatured, 200 Proof, Spectrum™ Chemical
SDP

Catalog Number 18600385
Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
64-17-5, 67-56-1, 67-63-0
C2H6O
46.07
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
CCO
64-17-5, 67-56-1, 67-63-0
C2H6O
46.07
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
CCO

A1038, 64-17-5, C2H5OH

Spectrum™ Chemical Alcohol, Anhydrous, Denatured, 200 Proof is ethanol that has added additives that render it undrinkable, bad tasting or even poisonous to discourage people from consuming it. The additives do not chemically change the alcohol and thus it is identical to a normal alcohol apart from it′s smell, color and taste and can be used for the exact same purposes as normal alcohol. Typical denaturants include methyl alcohol, isopropyl alcohol and methyl isobutyl ketone.
CAS Max % 84%, 5%, 10%
UN Number UN1987
Grade Reagent
Quantity 1 L
Packaging Poly Bottle
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit