Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol, Anhydrous, Reagent, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
19L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth Polyethylene Bottle
Poly Pail

Chemical Identifiers

67-56-1
CH4O
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
CHEBI:17790

High quality chemicals for laboratory and specialized industrial use

    For Histological Use
SMILES CO
IUPAC Name methanol
Packaging Aluminum Bottle
Specific Gravity 1L = 0.79kg
Flash Point 13.00°C
Quantity 1L
Chemical Name or Material Alcohol, Anhydrous, Reagent
Grade Baker Analyzed™ ACS Reagent