Learn More
Cell Biolabs IncSupplier Diversity Partner ALCOHOL ASSAY KIT COLOR  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA620
The Alcohol Assay Kit measures primary alcohols by an enzymatic, oxidation reaction, producing hydrogen peroxide which reacts with the kit's probe. Detection by colorimetric plate reader. 100 assays/kit including standards and unknowns.
Catalog No. 50-673-659