Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alcohol dehydrogenase 5 Mouse anti-Human, DyLight 550, Clone: OTI5G4, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270096R
Description
alcohol dehydrogenase 5 Monoclonal antibody specifically detects alcohol dehydrogenase 5 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
alcohol dehydrogenase 5 | |
OTI5G4 | |
Western Blot, Immunohistochemistry | |
Affinity Purified, Conjugated | |
ADH5 | |
Recombinant protein fragment corresponding to amino acids 1-266 of human alcohol dehydrogenase 5 (NP_000662) produced in E.coli. | |
Immunogen affinity purified | |
Core ESC Like Genes, Stem Cell Markers | |
Primary | |
128.0 |
Immunohistochemistry, Western Blot | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ADH-3, ADHXEC 1.1.1.1, alcohol dehydrogenase (class III), chi polypeptide, Alcohol dehydrogenase 5, alcohol dehydrogenase 5 (class III), chi polypeptide, Alcohol dehydrogenase class chi chain, alcohol dehydrogenase class-3, Alcohol dehydrogenase class-III, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.284, FALDH, FDHGSNOR, formaldehyde dehydrogenase, Glutathione-dependent formaldehyde dehydrogenase, GSH-FDH, S-(hydroxymethyl)glutathione dehydrogenase | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title