Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol Dehydrogenase, yeast, Affymetrix/USB™

Click to view available options
Quantity:
75kU
400kU
9031-72-5
MFCD00081305
9031-72-5
MFCD00081305
Product Type Alcohol Dehydrogenase
Format Lyophilized Powder
Concentration ≥300U/mg
Source Yeast
pH 8.8
Quantity 75kU
Storage Requirements -20°C
Physical Form Lyophilized powder
Chemical Name or Material Alcohol Dehydrogenase, yeast

EINECSNumber : 232-870-4