Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol-Phenol TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
100 mL
1 L
Packaging:
Amber Glass Bottle
108-95-2,64-17-5
C6H6O
94.11
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
phenol
OC1=CC=CC=C1
108-95-2,64-17-5
C6H6O
94.11
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
phenol
OC1=CC=CC=C1

A-293, 108-95-2

Spectrum™ Alcohol-Phenol TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
CAS Max % 0.999,0.001
UN Number UN1170
Packaging Amber Glass Bottle
Quantity 100 mL