Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol SDA 3A 200 Proof, Reagents

Catalog No. p-200020872
Click to view available options
Quantity:
20 L
55 gal.

Alcohol SDA 3A 200 Proof, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

Synonym Ethanol, Ethyl alcohol, Methylcarbinol

Specifications

Melting Point -114°C
CAS 67-56-1
Color Colorless
Boiling Point 78°C
Quantity 20 L
Packaging HDPE Winpak
Physical Form Liquid
Specific Gravity 0.7908
Molecular Formula CH4O
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Percent Purity Min. 99.5%
Grade Reagent
Chemical Name or Material Alcohol SDA 3A 200 Proof
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.