Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alconox™ Citrajet™ Low-Foaming Liquid Acid Cleaner

Liquid cleaner and metal brightener for removal of metal oxides, scales, salts and inorganic residues

Manufacturer:  Alconox™ 2030

 View more versions of this product

Catalog No. 16-000-148


Add to cart

Description

Description

Liquid cleaner and metal brightener for removal of metal oxides, scales, salts and inorganic residues

 

  • Concentrated, biodegradable and phosphate-free
  • Contains blend of organic acids and surfactants
  • Corrosion inhibited and free rinsing leaving no interfering residues
  • Ideal for acid washing and rinsing
  • Dilute 1:100 for use (10mL in 1L water)
  • Use to pass cleaning validation tests for lab accreditation and plant inspection approval
  • USDA authorized for use in federally inspected meat and poultry plants
  • pH 2.5

 

Used to clean:
Laboratory ware, sampling apparatus, dairy equipment optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, metal forgings, stampings and castings, industrial parts, pipes, tanks, boilers and reactors.

Used to remove:
Scale, metal oxides, metal complexes, trace inorganics, milkstone, soil, grit, buffing compound, grime, grease, fats, oils, particulates, amphoteric peptides, deposits, chemical and solvents.

Surfaces cleaned:
Glass, metal, stainless steel, porcelain, ceramic, plastic, rubber and fiberglass. Can be used on soft metals such as copper, bronze, aluminum, zinc and magnesium if rinsed promptly.

Cleaning method:
Laboratory warewashers, dishwashers, washer-sterilizers, washer-sanitizers, cart washers, spray washers, electrocleaning, ultrasonic cleaners flow through clean-in-place and manual cleaning if proper skin and eye protection is used.

Shelf life: 2 years from date of manufacture

Order Info

3.8L (1 gal.) 1 gal. makes 189L (50 gal.) cleaning solution.

Specifications

Specifications

Liquid
113.5L (30 gal.)
Pale yellow
Low-Foaming Liquid Acid Cleaner
2.5
Store closed and upright in a cool dry place at 15° to 30°C.
Drum
Removal of metal oxides, scales, salts and inorganic residues in labware washers, parts washers, power-spray systems
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit