Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aldehyde Dehydrogenase 3-A1/ALDH3A1 Mouse anti-Human, PE/Atto594, Clone: OTI1B6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270152PEATT594

Catalog No. N270152P594


Add to cart

Description

Description

Aldehyde Dehydrogenase 3-A1/ALDH3A1 Monoclonal antibody specifically detects Aldehyde Dehydrogenase 3-A1/ALDH3A1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Aldehyde Dehydrogenase 3-A1/ALDH3A1
OTI1B6
Flow Cytometry
Affinity Purified, Conjugated
ALDH3A1
Full length recombinant protein of human Aldehyde Dehydrogenase 3-A1/ALDH3A1 (NP_000682) produced in HEK293T cells.
Immunogen affinity purified
Vision
Primary
218.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
Aldehyde dehydrogenase 3, aldehyde dehydrogenase 3 family, member A1, Aldehyde dehydrogenase family 3 member A1, aldehyde dehydrogenase isozyme 3, aldehyde dehydrogenase type III, ALDH3aldehyde dehydrogenase, dimeric NADP-preferring, ALDHIII, EC 1.2.1, EC 1.2.1.5, MGC10406, stomach aldehyde dehydrogenase
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit