Learn More
MilliporeSigma AldeRed ALDH Detection Assay, 1 kit
Supplier:  MilliporeSigma SCR150
Encompass
The AldeRed ALDH Detection Kit provides cancer and stem cell scientists with new capabilities for live cell isolation and characterization. The AldeRed reagent is a red-shifted fluorescent substrate for aldehyde dehydrogenase (ALDH), allowing cells to be identified and isolated by flow cytometry with concurrent use of green fluorescent cell lines, antibodies, transgenic animals and reporter assays.
Catalog No. NC1402262
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.