Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALDH18A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NBP183324
Description
ALDH18A1 Polyclonal specifically detects ALDH18A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ALDH18A1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
aldehyde dehydrogenase 18 family, member A1, Aldehyde dehydrogenase family 18 member A1, delta-1-pyrroline-5-carboxylate synthase, EC 2.1.2.11, EC 2.3.3.13, GSASdelta1-pyrroline-5-carboxlate synthetase, MGC117316, P5CSdelta-1-pyrroline-5-carboxylate synthetase, PYCSEC 1.2.1.41, pyrroline-5-carboxylate synthetase (glutamate gamma-semialdehyde synthetase) | |
Rabbit | |
Affinity Purified | |
RUO | |
5832 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.1mg/mL | |
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
P54886 | |
ALDH18A1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:SVTFGTKSRVGMGGMEAKVKAALWALQGGTSVVIANGTHPKVSGHVITDIVEGKKVGTFFSEVKPAGPTVEQQGEMARSGGRMLATLEPEQRAEIIHHLADLLTDQRD | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction