Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ALDH18A1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519392
Description
This antibody is predicted to react with bovine, canine, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
This gene is a member of the aldehyde dehydrogenase family and encodes a bifunctional ATP- and NADPH-dependent mitochondrial enzyme with both gamma-glutamyl kinase and gamma-glutamyl phosphate reductase activities. The encoded protein catalyzes the reduction of glutamate to delta1-pyrroline-5-carboxylate, a critical step in the de novo biosynthesis of proline, ornithine and arginine. Mutations in this gene lead to hyperammonemia, hypoornithinemia, hypocitrullinemia, hypoargininemia and hypoprolinemia and may be associated with neurodegeneration, cataracts and connective tissue diseases. Alternatively spliced transcript variants, encoding different isoforms, have been described for this gene.
Specifications
ALDH18A1 | |
Polyclonal | |
Unconjugated | |
ALDH18A1 | |
2810433K04Rik; ADCL3; AI429789; aldehyde dehydrogenase 18 family member A1; aldehyde dehydrogenase 18 family, member A1; aldehyde dehydrogenase 18A1; aldehyde dehydrogenase family 18 member A1; aldh18a1; ARCL3A; cb842; cb899; delta1-pyrroline-5-carboxlate synthetase; delta-1-pyrroline-5-carboxylate synthase; delta-1-pyrroline-5-carboxylate synthetase; Gamma-glutamyl kinase; Gamma-glutamyl phosphate reductase; GK; Glutamate 5-kinase; glutamate gamma-semialdehyde synthetase; Glutamate-5-semialdehyde dehydrogenase; Glutamyl-gamma-semialdehyde dehydrogenase; GPR; GSAS; MGC117316; P5CS; Pycs; pyrroline-5-carboxylate synthetase; pyrroline-5-carboxylate synthetase (glutamate gamma-semialdehyde synthetase); sb:cb881; similar to pyrroline-5-carboxylate synthetase isoform 1; SPG9A; SPG9B; wu:fa91f10; wu:fi05f11; wu:fi17d12 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
56454, 5832 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P54886, Q9Z110 | |
ALDH18A1 | |
Synthetic peptide sequence (SEHGSLKYLH) corresponding to the C-terminus amino acids of ALDH18A1 (aa 775-784). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction