Learn More
ALDH1A1 Rabbit anti-Human, KHL, Clone: 4J6, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB10994X25UL
 View more versions of this product
Catalog No. ZRB10994XMI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
ALDH1A1 Monoclonal antibody specifically detects ALDH1A1 in Human samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.
Specifications
Specifications
ALDH1A1
4J6
KHL
Purified
ALDH1A1, ALDC, ALDH1, PUMB1
KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal region of human Retinal dehydrogenase 1.
∼54 kDa observed; 54.86 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
4 x 25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Human
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_000680.2
Retinal dehydrogenase 1;RALDH 1;RalDH1;ALDH-E1;ALHDII;Aldehyde dehydrogenase family 1 member A1;Aldehyde dehydrogenase cytosolic
Rabbit
IgG
Protein A Purified
Neuroscience
Primary
4X