Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH1A3 Mouse anti-Human, PE-Cyanine7, Clone: OTI4E8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270154PECY7

Catalog No. N270154PEC7


Add to cart

Description

Description

ALDH1A3 Monoclonal antibody specifically detects ALDH1A3 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

ALDH1A3
OTI4E8
Flow Cytometry
Affinity Purified, Conjugated
ALDH1A3
Recombinant protein fragment corresponding to amino acids 1-100 and 413-512 of human ALDH1A3 (NP_000648) produced in E.coli.
Immunogen affinity purified
Vision
Primary
220.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
aldehyde dehydrogenase 1 family, member A3, Aldehyde dehydrogenase 6, aldehyde dehydrogenase family 1 member A3, ALDH1A6, ALDH6acetaldehyde dehydrogenase 6, EC 1.2.1, EC 1.2.1.5, RalDH3, RALDH-3, Retinaldehyde dehydrogenase 3
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit