Learn More
ALDH1L1 Mouse anti-Canine, Human, Clone: OTI5G8, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA501870
Catalog No. 50-167-2977
Add to cart
Description
Description
ALDH1L1 catalyzes the conversion of 10-formyltetrahydrofolate, NADP, and water to tetrahydrofolate, NADPH, and carbon dioxide. The encoded protein belongs to the aldehyde dehydrogenase family and is responsible for formate oxidation in vivo. Deficiencies in this gene can result in an accumulation of formate and subsequent methanol poisoning.
Specifications
Specifications
ALDH1L1
Monoclonal
Unconjugated
ALDH1L1
Liquid
ALDH1L1, LOC100855730
Full length human recombit protein of human ALDH1L1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI5G8
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O75891
10-FTHFDH, 10-fTHF, FDH, FTHFD
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100855730, 10840