Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALDH1L1 Antibody (OTI3D2), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271629AF700
Description
ALDH1L1 Monoclonal specifically detects ALDH1L1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ALDH1L1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ALDH1L1 | |
Full length recombinant protein of human ALDH1L1 (NP_036322) produced in HEK293T cells. | |
0.1 mL | |
10840.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3D2 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
10-formyltetrahydrofolate dehydrogenase, aldehyde dehydrogenase 1 family, member L1, aldehyde dehydrogenase family 1 member L1, Cytosolic 10-formyltetrahydrofolate dehydrogenase, DKFZp781N0997, EC 1.5.1.6,10-FTHFDH, FDH, formyltetrahydrofolate dehydrogenase, FTHFD10-fTHF | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction