Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH1L2 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548161

Catalog No. PA548161

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with xenopus based on sequence homology.

This gene encodes a member of both the aldehyde dehydrogenase superfamily and the formyl transferase superfamily. This member is the mitochondrial form of 10-formyltetrahydrofolate dehydrogenase (FDH), which converts 10-formyltetrahydrofolate to tetrahydrofolate and CO2 in an NADP(+)-dependent reaction, and plays an essential role in the distribution of one-carbon groups between the cytosolic and mitochondrial compartments of the cell. Alternatively spliced transcript variants have been found for this gene.[provided by RefSeq, Oct 2010]
Specifications

Specifications

ALDH1L2
Polyclonal
Unconjugated
Aldh1l2
Liquid
ALDH1L2
KLH conjugated synthetic peptide between 743-772 amino acids from the C-terminal region of human ALDH1L2.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide
Q8K009
Mitochondrial 10-formyltetrahydrofolate dehydrogenase, Mitochondrial 10-FTHFDH, mtFDH, Aldehyde dehydrogenase family 1 member L2, ALDH1L2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
216188
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit