Learn More
ALDH2 Antibody (OTI4H2), DyLight 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270151UV
Catalog No. NBP270151UV
Add to Cart
Description
Description
ALDH2 Monoclonal specifically detects ALDH2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
ALDH2
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
ALDH2
Full length recombinant protein of human ALDH2 (NP_000681) produced in HEK293T cells.
0.1 mL
217.0
Store at 4°C in the dark.
Flow Cytometry, Immunocytochemistry, Immunofluorescence
OTI4H2
Flow Cytometry, Immunocytochemistry/Immunofluorescence
acetaldehyde dehydrogenase 2, aldehyde dehydrogenase 2 family (mitochondrial), aldehyde dehydrogenase, mitochondrial, ALDH class 2, ALDH-E2, ALDHI, ALDM, EC 1.2.1, EC 1.2.1.3, liver mitochondrial ALDH, MGC1806, nucleus-encoded mitochondrial aldehyde dehydrogenase 2
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG3
Recommendations
Recommendations