Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALDH2 Mouse anti-Human, FITC, Clone: OTI4H2, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270151F
Description
ALDH2 Monoclonal antibody specifically detects ALDH2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence.Specifications
ALDH2 | |
OTI4H2 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
Affinity Purified, Conjugated | |
ALDH2 | |
Full length recombinant protein of human ALDH2 (NP_000681) produced in HEK293T cells. | |
Immunogen affinity purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
FITC | |
PBS with 0.05% Sodium Azide | |
acetaldehyde dehydrogenase 2, aldehyde dehydrogenase 2 family (mitochondrial), aldehyde dehydrogenase, mitochondrial, ALDH class 2, ALDH-E2, ALDHI, ALDM, EC 1.2.1, EC 1.2.1.3, liver mitochondrial ALDH, MGC1806, nucleus-encoded mitochondrial aldehyde dehydrogenase 2 | |
Mouse | |
IgG3 | |
0.1 mL | |
Primary | |
217.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title