Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH2 Mouse anti-Human, PE/Cy5.5, Clone: OTI4H2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270151PECY55

Catalog No. N270151PEC5


Add to cart

Description

Description

ALDH2 Monoclonal antibody specifically detects ALDH2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

ALDH2
OTI4H2
Flow Cytometry
Affinity Purified, Conjugated
ALDH2
Full length recombinant protein of human ALDH2 (NP_000681) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
acetaldehyde dehydrogenase 2, aldehyde dehydrogenase 2 family (mitochondrial), aldehyde dehydrogenase, mitochondrial, ALDH class 2, ALDH-E2, ALDHI, ALDM, EC 1.2.1, EC 1.2.1.3, liver mitochondrial ALDH, MGC1806, nucleus-encoded mitochondrial aldehyde dehydrogenase 2
Mouse
IgG3
0.1 mL
Primary
217.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit