Learn More
ALDH3A2 Mouse anti-Canine, Human, Non-human primate, Rat, Clone: OTI2D3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525636
Catalog No. PIMA525636
Add to cart
Description
Description
Aldh3A2 is a member of the aldehyde dehydrogenase superfamily, a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of aliphatic and aromatic aldehydes. Aldehyde dehydrogenase enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. Aldh3A2 catalyzes the oxidation of long-chain aliphatic aldehydes to fatty acids. Mutations in the Aldh3A2 gene cause Sjogren-Larrson syndrome, an inherited neurocutaneous disorder. Patients with this disorder display ichthyosis, mental retardation and spastic diplegia. The pathogenesis of the cutaneous and neurological symptoms is thought to result from abnormal lipid accumulation in the membranes of skin and brain, the formation of aldehyde Schiff base adducts with amine-containing lipids or proteins, or defective eicosanoid metabolism.
Specifications
Specifications
ALDH3A2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q60HH8, P30839, P51648
Aldh3a2; aldehyde dehydrogenase 3 family, member A2; Aldh4; FALDH; alcohol dehydrogenase family 3, subfamily A2; aldehyde dehydrogenase 3A2; aldehyde dehydrogenase 4; aldehyde dehydrogenase family 3 member A2; aldehyde dehydrogenase family 3, subfamily A2; fatty aldehyde dehydrogenase; microsomal aldehyde dehydrogenase; msALDH
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100688308, 102138923, 224, 65183
Western Blot
OTI2D3
Unconjugated
ALDH3A2
Liquid
ALDH3A2
Full length human recombit protein of ALDH3A2 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Non-human primate, Rat