Learn More
ALDH3B1 Mouse anti-Human, Clone: OTI2F6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811463

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The aldehyde dehydrogenases are a family of isozymes that may play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. This particular gene spans about 20 kb of genomic DNA and is composed of 9 coding exons. The gene is highly expressed in kidney and lung. The functional significance of this gene as well as the cellular localization of its product are presently unknown. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
ALDH3B1 | |
Monoclonal | |
Unconjugated | |
ALDH3B1 | |
ALDH4, ALDH7 | |
Mouse | |
Affinity Chromatography | |
RUO | |
221 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2F6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P43353 | |
ALDH3B1 | |
Human recombit protein fragment corresponding to amino acids 1-465 of human ALDH3B1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.