Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH3B2 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen ALDH3B2
Applications Immunohistochemistry
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB096191
View Documents
Novus Biologicals
NBP304930100UL
100 μg Each for $320.71
Add to cart
 
NB096192
View Documents
Novus Biologicals
NBP30493020UL
20 μg Each for $138.37
Add to cart
 
Description

Description

ALDH3B2 Polyclonal antibody specifically detects ALDH3B2 in Rat samples. It is validated for Immunohistochemistry
Specifications

Specifications

ALDH3B2
Unconjugated
Rabbit
Rat
acetaldehyde dehydrogenase 8, aldehyde dehydrogenase 3 family, member B2, Aldehyde dehydrogenase 8, ALDH8aldehyde dehydrogenase family 3 member B2, EC 1.2.1, EC 1.2.1.5
Recombinant fusion protein containing a sequence corresponding to amino acids 296-385 of human ALDH3B2 (NP_000686.2).
Polyclonal
Affinity purified
Immunohistochemistry
Purified
RUO
PBS with 50% glycerol, pH7.3.
222
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit