Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ ALDH6A1 Human Recombinant Protein
Highly purified. Generating reliable and reproducible results.
$344.00 - $976.00
Specifications
Concentration | 0.25mg/mL |
---|---|
For Use With (Application) | SDS-Page |
Formulation | Phosphate buffered saline (pH7.4) containing 10% glycerol, 1mM DTT |
Gene ID (Entrez) | 4329 |
Molecular Weight | TMW: 56.8kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2520902
|
Novus Biologicals™
NBP2520900.02MG |
0.02mg |
Each for $344.00
|
|
NBP25209001
|
Novus Biologicals™
NBP2520900.1MG |
0.1mg |
Each for $976.00
|
|
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 34-535 of Human ALDH6A1 The Recombinant Human ALDH6A1 Protein is derived from E. coli. The Recombinant Human ALDH6A1 Protein has been validated for the following applications: SDS-Page.Specifications
0.25mg/mL | |
Phosphate buffered saline (pH7.4) containing 10% glycerol, 1mM DTT | |
TMW: 56.8kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
ALDH6A1 | |
Yes |
SDS-Page | |
4329 | |
>85%, by SDS-PAGE | |
aldehyde dehydrogenase 6 family, member A1, Aldehyde dehydrogenase family 6 member A1, EC 1.2.1.18, EC 1.2.1.27, malonate-semialdehyde dehydrogenase, Malonate-semialdehyde dehydrogenase [acylating], methylmalonate-semialdehyde dehydrogenase [acylating], mitochondrial, MGC40271, MMSADHA, MMSDHmitochondrial acylating methylmalonate-semialdehyde dehydrogenase | |
Unlabeled |
For research use only and is not approved for use in humans or in clinical diagnosis
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title