Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

ALDH6A1 Mouse anti-Human, Clone: 147CT8.3.4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524714

Catalog No. PIMA524714



Description

Description

This protein belongs to the aldehyde dehydrogenases family of proteins. This enzyme plays a role in the valine and pyrimidine catabolic pathways. The product of this gene, a mitochondrial methylmalonate semialdehyde dehydrogenase, catalyzes the irreversible oxidative decarboxylation of malonate and methylmalonate semialdehydes to acetyl- and propionyl-CoA. Methylmalonate semialdehyde dehydrogenase deficiency is characterized by elevated beta-alanine, 3-hydroxypropionic acid, and both isomers of 3-amino and 3-hydroxyisobutyric acids in urine organic acids.
Specifications

Specifications

ALDH6A1
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide
Q02252
Methylmalonate-semialdehyde dehydrogenase [acylating], mitochondrial, MMSDH, Malonate-semialdehyde dehydrogenase [acylating], Aldehyde dehydrogenase family 6 member A1, ALDH6A1, MMSDH
Mouse
IgG1, kappa
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4329
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
147CT8.3.4
Unconjugated
ALDH6A1
Liquid
ALDH6A1
human ALDH6A1 recombit protein
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit