Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH7A1 Rabbit anti-Human, DyLight 488, Clone: 101, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289926G

Catalog No. NB110138


Add to cart

Description

Description

ALDH7A1 Monoclonal specifically detects ALDH7A1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

ALDH7A1
101
50 mM Sodium Borate
aldehyde dehydrogenase 7 family, member A1, Aldehyde dehydrogenase family 7 member A1,26g turgor protein homolog, Alpha-AASA dehydrogenase, ATQ1Antiquitin-1, Betaine aldehyde dehydrogenase, Delta1-piperideine-6-carboxylate dehydrogenase, EC 1.2.1, EC 1.2.1.3, EC 1.2.1.31, EC 1.2.1.8, EPDalpha-aminoadipic semialdehyde dehydrogenase, FLJ11738, FLJ92814, P6c dehydrogenase, PDEantiquitin-1
This antibody was obtained from a rabbit immunized with purified, recombinant Human ALDH7A1 (P49419-2; Ser2-Gln511).
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunofluorescence
DyLight 488
Purified
Rabbit
IgG
0.1 mL
Core ESC Like Genes, Stem Cell Markers
Primary
501
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit