Learn More
Abnova™ ALDH8A1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00064577T01
Catalog No. 89-933-126
Add to cart
Description
Description
Specifications
Specifications
293T
NM_022568.2
pCMV-ALDH8A1 full-length
100 μL
Human
ALDH8A1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 53.68kDa
Transient overexpression cell lysate was tested with anti-ALDH8A1 antibody (Mfr. No. H00064577-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot