Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDOC Mouse anti-Human, Clone: 859CT9.5.3, Abnova™

Mouse monoclonal antibody raised against recombinant human ALDOC.

Manufacturer:  Abnova Corporation MAB12876

Catalog No. 89-933-551


Add to cart

Description

Description

This gene encodes a member of the class I fructose-biphosphate aldolase gene family. Expressed specifically in the hippocampus and Purkinje cells of the brain, the encoded protein is a glycolytic enzyme that catalyzes the reversible aldol cleavage of fructose-1,6-biphosphate and fructose 1-phosphate to dihydroxyacetone phosphate and either glyceraldehyde-3-phosphate or glyceraldehyde, respectively. [provided by RefSeq]

Specifications

Specifications

ALDOC
859CT9.5.3
Mouse monoclonal antibody raised against recombinant human ALDOC.
In ascites (0.09% sodium azide).
ALDC
Mouse
IgG3
RUO
Primary
230
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot (1:5000) The optimal working dilution should be determined by the end user.
Liquid
ALDOC
Recombinant His fusion protein corresponding to human ALDOC.
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit