Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aldolase A Rabbit anti-Human, Mouse, Rat, Zebrafish, Clone: JM54-19, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275402
Description
Aldolase A Monoclonal specifically detects Aldolase A in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Aldolase A | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
ALDA, aldolase A, fructose-bisphosphate, EC 4.1.2.13, fructose-1,6-bisphosphate triosephosphate-lyase, fructose-bisphosphate aldolase A, GSD12, Lung cancer antigen NY-LU-1, MGC10942, MGC17716, MGC17767, Muscle-type aldolase | |
Rabbit | |
39 kDa | |
100 μL | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
JM54-19 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
ALDOA | |
Synthetic peptide within N-terminal human Aldolase A . (SwissProt: P04075 Human; SwissProt: P05064 Mouse; SwissProt: P05065 Rat) | |
Protein A purified | |
Cancer, Cell Cycle and Replication | |
226.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction