Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aldolase B Mouse anti-Human, Allophycocyanin/Cy7, Clone: OTI3C6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270162APCCY7

Catalog No. N270162APC7


Add to cart

Description

Description

Aldolase B Monoclonal antibody specifically detects Aldolase B in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Aldolase B
OTI3C6
Flow Cytometry
Affinity Purified, Conjugated
ALDOB
Full length recombinant protein of human Aldolase B (NP_000026) produced in HEK293 cell.
Immunogen affinity purified
Cancer
Primary
229.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
ALDO2, aldolase B, fructose-bisphosphate, EC 4.1.2.13, fructose-bisphosphatase, fructose-bisphosphate aldolase B
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit